ENA053246125 Enamine

1-[4-(propan-2-yl)phenyl]-1H-imidazole-2-thiol

  • MDL number MFCD06382006
  • Cas Number : 358619-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-144
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12053
Availability details for
Quantity
  • Availability
  • Add to Cart