ENA053246130 Enamine

{[1-(2,5-dimethylphenyl)-1H-imidazol-2-yl]thio}acetic acid

  • MDL number MFCD06380397
  • Cas Number : 851398-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12058
Availability details for
Quantity
  • Availability
  • Add to Cart