ENA053246135 Enamine

N-{2-[4-(2-chloroacetyl)phenyl]ethyl}acetamide

  • MDL number MFCD06380393
  • Cas Number : 63077-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12063
Availability details for
Quantity
  • Availability
  • Add to Cart