ENA053246139 Enamine

2-(4-methyl-2-oxo-2,3-dihydro-1,3-thiazol-3-yl)acetic acid

  • MDL number MFCD06380390
  • Cas Number : 68104-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12067
Availability details for
Quantity
  • Availability
  • Add to Cart