ENA053246149 Enamine

N1,N1-diethyl-N4-[(thiophen-2-yl)methyl]benzene-1,4-diamine

  • MDL number MFCD04525945
  • Cas Number : 851457-29-9
  • Molecular Weight : 260.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12077
Availability details for
Quantity
  • Availability
  • Add to Cart