ENA053246151 Enamine

2,5-dimethyl-1-[1-(propan-2-yl)-1H-pyrazol-5-yl]-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD06382016
  • Cas Number : 956386-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12079
Availability details for
Quantity
  • Availability
  • Add to Cart