ENA053246159 Enamine

1-butyl-2-(4-methoxyphenyl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD06382827
  • Cas Number : 851721-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12087
Availability details for
Quantity
  • Availability
  • Add to Cart