ENA053246169 Enamine

6-(2-amino-1,3-thiazol-4-yl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD00828995
  • Cas Number : 105316-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12097
Availability details for
Quantity
  • Availability
  • Add to Cart