ENA053246179 Enamine

6-cyclopropyl-1-(1,1-dioxidotetrahydrothien-3-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06382834
  • Cas Number : 851721-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12107

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12107
Availability details for
Quantity
  • Availability
  • Add to Cart