ENA053246189 Enamine

1-(2,3-dimethylphenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD05625734
  • Cas Number : 851721-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12117
Availability details for
Quantity
  • Availability
  • Add to Cart