ENA053246190 Enamine

1-(3,5-dimethylphenyl)-1H-tetrazole-5-thiol

  • MDL number MFCD06382857
  • Cas Number : 851721-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12118
Availability details for
Quantity
  • Availability
  • Add to Cart