ENA053246191 Enamine

1-(3-chlorophenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD01249730
  • Cas Number : 22384-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12119
Availability details for
Quantity
  • Availability
  • Add to Cart