ENA053246195 Enamine

2-({1-[4-(propan-2-yl)phenyl]-1H-imidazol-2-yl}sulfanyl)acetic acid

  • MDL number MFCD06382856
  • Cas Number : 851721-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12123
Availability details for
Quantity
  • Availability
  • Add to Cart