ENA053246202 Enamine

3-{5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl}propan-1-ol

  • MDL number MFCD04967214
  • Cas Number : 832737-20-9
  • Molecular Weight : 206.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12130
Availability details for
Quantity
  • Availability
  • Add to Cart