ENA053246209 Enamine

4-(4-bromophenyl)-5-(pyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03722487
  • Cas Number : 127055-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12137
Availability details for
Quantity
  • Availability
  • Add to Cart