ENA053246211 Enamine

2-{[(dimethylcarbamoyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD06385598
  • Cas Number : 851788-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12139
Availability details for
Quantity
  • Availability
  • Add to Cart