ENA053246213 Enamine

2-bromo-1-[4-(difluoromethoxy)-3-methoxyphenyl]ethanone

  • MDL number MFCD06382865
  • Cas Number : 149169-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12141

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12141
Availability details for
Quantity
  • Availability
  • Add to Cart