ENA053246219 Enamine

2-chloro-N-[(furan-2-yl)methyl]-N-methylacetamide

  • MDL number MFCD06655259
  • Cas Number : 75228-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12147
Availability details for
Quantity
  • Availability
  • Add to Cart