ENA053246222 Enamine

(2E)-3-{4-[(3,5-dimethylisoxazol-4-yl)methoxy]-3-methoxyphenyl}acrylic acid

  • MDL number MFCD06655260
  • Cas Number : 851903-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12150
Availability details for
Quantity
  • Availability
  • Add to Cart