ENA053246223 Enamine

N-{4-[(4-bromophenyl)thio]phenyl}-2-chloroacetamide

  • MDL number MFCD00665609
  • Cas Number : 38007-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12151
Availability details for
Quantity
  • Availability
  • Add to Cart