ENA053246226 Enamine

2-[(2,5-dimethylphenyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD06368757
  • Cas Number : 795290-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12154
Availability details for
Quantity
  • Availability
  • Add to Cart