ENA053246228 Enamine

1-(1,1-dioxo-1lambda6,2-benzothiazol-3-yl)piperidine-4-carboxylic acid

  • MDL number MFCD06380386
  • Cas Number : 851170-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12156
Availability details for
Quantity
  • Availability
  • Add to Cart