ENA053246234 Enamine

1-(3-aminopropyl)azepan-2-one

  • MDL number MFCD00036060
  • Cas Number : 24566-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...175
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12162

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-12162
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart