ENA053246239 Enamine

1,6-bis(propan-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06380320
  • Cas Number : 851398-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12167
Availability details for
Quantity
  • Availability
  • Add to Cart