ENA053246244 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]-2-methylpropanamide

  • MDL number MFCD06382881
  • Cas Number : 851722-10-6
  • Molecular Weight : 261.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12172
Availability details for
Quantity
  • Availability
  • Add to Cart