ENA053246248 Enamine

5-acetyl-2-mercapto-6-phenylnicotinonitrile

  • MDL number MFCD06382860
  • Cas Number : 851722-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12176

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12176
Availability details for
Quantity
  • Availability
  • Add to Cart