ENA053246251 Enamine

ethyl 2-(chloromethyl)-6-methyl-4-oxo-3H,4H-furo[2,3-d]pyrimidine-5-carboxylate

  • MDL number MFCD06382868
  • Cas Number : 851722-05-9
  • Molecular Weight : 270.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12179
Availability details for
Quantity
  • Availability
  • Add to Cart