ENA053246261 Enamine

2-chloro-N-(2,3-dihydro-1,4-benzodioxin-6-yl)-N-[(thiophen-2-yl)methyl]acetamide

  • MDL number MFCD06382867
  • Cas Number : 851722-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12189
Availability details for
Quantity
  • Availability
  • Add to Cart