ENA053246262 Enamine

2-amino-5-cyano-N-phenyl-6-sulfanylpyridine-3-carboxamide

  • MDL number MFCD06382861
  • Cas Number : 496844-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12190
Availability details for
Quantity
  • Availability
  • Add to Cart