ENA053246274 Enamine

8-methyl-1,3,8-triazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD01679435
  • Cas Number : 52094-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283...285
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12202

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12202
Availability details for
Quantity
  • Availability
  • Add to Cart