ENA053246277 Enamine

2-chloro-N-methyl-N-(1,2,3,4-tetrahydronaphthalen-1-yl)acetamide

  • MDL number MFCD06384312
  • Cas Number : 276886-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12205
Availability details for
Quantity
  • Availability
  • Add to Cart