ENA053246278 Enamine

2-{[(4,5-dimethoxy-2-methylphenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD06384972
  • Cas Number : 850020-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12206
Availability details for
Quantity
  • Availability
  • Add to Cart