ENA053246284 Enamine

5-phenyl-7-(trifluoromethyl)-[1,3]thiazolo[4,5-b]pyridin-2-amine

  • MDL number MFCD06385033
  • Cas Number : 850021-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12212
Availability details for
Quantity
  • Availability
  • Add to Cart