ENA053246288 Enamine

(2,7-dimethyl-4-oxo-5,6,7,8-tetrahydro[1]benzothieno[2,3-d]pyrimidin-3(4H)-yl)acetic acid

  • MDL number MFCD06385038
  • Cas Number : 850021-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12216

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12216
Availability details for
Quantity
  • Availability
  • Add to Cart