ENA053246302 Enamine

ethyl 2-amino-5-cyano-6-sulfanylpyridine-3-carboxylate

  • MDL number MFCD06386706
  • Cas Number : 625370-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12230
Availability details for
Quantity
  • Availability
  • Add to Cart