ENA053246312 Enamine

1-oxo-3-phenyl-3,4-dihydro-1H-2-benzopyran-6-carboxylic acid

  • MDL number MFCD06385044
  • Cas Number : 850021-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-231
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12240
Availability details for
Quantity
  • Availability
  • Add to Cart