ENA053246315 Enamine

5-(4-fluorophenyl)furan-2-carboxylic acid

  • MDL number MFCD02104577
  • Cas Number : 73269-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-202
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12243
Availability details for
Quantity
  • Availability
  • Add to Cart