ENA053246319 Enamine

2-(2,2-dimethylpropanamido)acetic acid

  • MDL number MFCD06385042
  • Cas Number : 23891-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12247
Availability details for
Quantity
  • Availability
  • Add to Cart