ENA053246334 Enamine

4-(5-methylthiophen-2-yl)-4-oxobutanoic acid

  • MDL number MFCD06201682
  • Cas Number : 22988-52-9
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 87-88
Iupac Name 4-(5-methyl-2-thienyl)-4-oxobutanoic acid
Inchi Code 1S/C9H10O3S/c1-6-2-4-8(13-6)7(10)3-5-9(11)12/h2,4H,3,5H2,1H3,(H,11,12)
Inchi Key BBGWOJJYYCDOAN-UHFFFAOYSA-N

Description

Melting Point 87-88
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12262
Availability details for
Quantity
  • Availability
  • Add to Cart