ENA053246336 Enamine

N-(4-methylphenyl)-2-(piperazin-1-yl)acetamide

  • MDL number MFCD06386688
  • Cas Number : 89473-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12264
Availability details for
Quantity
  • Availability
  • Add to Cart