ENA053246343 Enamine

[1-benzyl-3-(3-methoxyphenyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD06385606
  • Cas Number : 956573-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12271
Availability details for
Quantity
  • Availability
  • Add to Cart