ENA053246349 Enamine

(4-chlorophenyl)(cyclopropyl)methanamine

  • MDL number MFCD05191965
  • Cas Number : 123312-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12277
Availability details for
Quantity
  • Availability
  • Add to Cart