ENA053246354 Enamine

6-oxo-5-(4-phenyl-1,3-thiazol-2-yl)-2-sulfanyl-1,6-dihydropyridine-3-carbonitrile

  • MDL number MFCD06655271
  • Cas Number : 851879-14-6
  • Molecular Weight : 311.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12282
Availability details for
Quantity
  • Availability
  • Add to Cart