ENA053246357 Enamine

5-(3-chlorophenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06655273
  • Cas Number : 41491-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-179
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12285
Availability details for
Quantity
  • Availability
  • Add to Cart