ENA053246358 Enamine

5-(4-ethoxyphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06655274
  • Cas Number : 84159-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12286
Availability details for
Quantity
  • Availability
  • Add to Cart