ENA053246361 Enamine

4-amino-5-(2-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655276
  • Cas Number : 851879-31-7
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12289
Availability details for
Quantity
  • Availability
  • Add to Cart