ENA053246362 Enamine

4-amino-5-(2,4-dimethoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655277
  • Cas Number : 503004-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12290
Availability details for
Quantity
  • Availability
  • Add to Cart