ENA053246363 Enamine

4-amino-5-(4-fluorobenzyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655278
  • Cas Number : 151297-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12291
Availability details for
Quantity
  • Availability
  • Add to Cart