ENA053246365 Enamine

2-[2-(3-bromophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06655279
  • Cas Number : 851879-29-3
  • Molecular Weight : 298.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-105
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12293
Availability details for
Quantity
  • Availability
  • Add to Cart