ENA053246366 Enamine

2-[2-(2,3-dimethoxyphenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06655280
  • Cas Number : 851879-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12294
Availability details for
Quantity
  • Availability
  • Add to Cart